Product Image
Thermo Scientific Chemicals

Mephenytoin

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Catalog number 466690250
Price (EUR)
133,00
Each
Quantity:
25 mg
Request bulk or custom format
Price (EUR)
133,00
Each
Chemical Identifiers
CAS50-12-4
IUPAC Name5-ethyl-3-methyl-5-phenylimidazolidine-2,4-dione
Molecular FormulaC12H14N2O2
InChI KeyGMHKMTDVRCWUDX-UHFFFAOYNA-N
SMILESCCC1(NC(=O)N(C)C1=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=98.0 %
Appearance (Form)crystalline powder
Appearance (Color)White to almost white
Water=<2.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only