Panobinostat
Thermo Scientific Chemicals

Panobinostat

CAS: 404950-80-7 | C21H23N3O2, C21H23N3O2 | 349.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 467000010
Price (EUR)
520,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
520,00
Each
Chemical Identifiers
CAS404950-80-7
IUPAC NameN-hydroxy-3-[4-({[2-(2-methyl-1H-indol-3-yl)ethyl]amino}methyl)phenyl]prop-2-enamide
Molecular FormulaC21H23N3O2
InChI KeyFPOHNWQLNRZRFC-UHFFFAOYSA-N
SMILESCC1=C(CCNCC2=CC=C(C=CC(=O)NO)C=C2)C2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Almost white to yellow to beige to light brown
Loss on drying=<6 % (1 g, 105°C, 3 h)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only