Besifloxacin hydrochloride
Thermo Scientific Chemicals

Besifloxacin hydrochloride

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
467501000100 mg
467505000500 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 467501000
Price (EUR)
109,00
Each
Quantity:
100 mg
Request bulk or custom format
Price (EUR)
109,00
Each
Chemical Identifiers
CAS405165-61-9
IUPAC Name7-(3-aminoazepan-1-yl)-8-chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride
Molecular FormulaC19H22Cl2FN3O3
InChI KeyPMQBICKXAAKXAY-UHFFFAOYNA-N
SMILESCl.NC1CCCCN(C1)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=C1Cl)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration Argentometric97.5 to 102.5 %
Appearance (Color)White to yellow to light brown
HPLC>=97.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only