Carboxine
Thermo Scientific Chemicals

Carboxine

CAS: 5234-68-4 | C12H13NO2S, C12H13NO2S | 235.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 467620010
Price (EUR)
49,40
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
49,40
Each
Chemical Identifiers
CAS5234-68-4
IUPAC Name2-methyl-N-phenyl-5,6-dihydro-1,4-oxathiine-3-carboxamide
Molecular FormulaC12H13NO2S
InChI KeyGYSSRZJIHXQEHQ-UHFFFAOYSA-N
SMILESCC1=C(SCCO1)C(=O)NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to light beige
Appearance (Form)Powder or crystals
Melting point88°C to 93°C
Water=<1 %
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only