2-Bromoanthraquinone, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromoanthraquinone, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 467790050
Price (EUR)
94,30
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
94,30
Each
Chemical Identifiers
CAS572-83-8
IUPAC Name2-bromo-9,10-dihydroanthracene-9,10-dione
Molecular FormulaC14H7BrO2
InChI KeyVTSDGYDTWADUJQ-UHFFFAOYSA-N
SMILESBrC1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Light yellow to brown to dark green
HPLC>=97.5 %
Appearance (Form)Crystalline powder and chunks
Melting point205°C to 211°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only