Pictilisib, 95%
Thermo Scientific Chemicals

Pictilisib, 95%

CAS: 957054-30-7 | C23H27N7O3S2, C23H27N7O3S2 | 513.64 g/mol
Have Questions?
Catalog NumberQuantity
467861000100 mg
Catalog number 467861000
Price (EUR)
-
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS957054-30-7
IUPAC Name4-{6-[(4-methanesulfonylpiperazin-1-yl)methyl]-4-(morpholin-4-yl)thieno[3,2-d]pyrimidin-2-yl}-1H-indazole
Molecular FormulaC23H27N7O3S2
InChI KeyLHNIIDJUOCFXAP-UHFFFAOYSA-N
SMILESCS(=O)(=O)N1CCN(CC2=CC3=NC(=NC(N4CCOCC4)=C3S2)C2=C3C=NNC3=CC=C2)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
MScomplies
NMR (1H-NMR)Consistent with structure
Appearance (Form)Powder or solid
HPLC>=94.0 %
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only