T0901317, 95%
Thermo Scientific Chemicals

T0901317, 95%

CAS: 293754-55-9 | C17H12F9NO3S, C17H12F9NO3S | 481.33 g/mol
Have Questions?
Catalog NumberQuantity
467871000100 mg
Catalog number 467871000
Price (EUR)
-
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS293754-55-9
IUPAC NameN-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]-N-(2,2,2-trifluoroethyl)benzenesulfonamide
Molecular FormulaC17H12F9NO3S
InChI KeySGIWFELWJPNFDH-UHFFFAOYSA-N
SMILESOC(C1=CC=C(C=C1)N(CC(F)(F)F)S(=O)(=O)C1=CC=CC=C1)(C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=94.0 %
MScomplies
NMR (1H-NMR)Consistent with structure
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystalline powder or solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only