Search
| Catalog Number | Description |
|---|---|
| 50003135 | ASTar™ instrument system |
| 50003905 | ASTar™ Consumable Kit |
| 50003855 | ASTar™ Frozen Insert |
Achieve comprehensive antimicrobial sensitivity testing (AST) and reporting from positive blood culture within ∼6 hours with fully automated Q-linea ASTar™ System.
Proactively manage infection in critically ill patients by gaining early antimicrobial susceptibility data that supports timely and effective stewardship efforts.
Support the delivery of optimized treatment decisions by providing sensitivity testing results covering a broad range of antimicrobials, for both fastidious and non-fastidious pathogens, using the ASTar™ System.
The ASTar Instrument is an in vitro diagnostic medical device for performing quantitative determination of antimicrobial susceptibility.
Test results are an aid in the determination of antimicrobial agents of choice to treat patients with bacteremia in conjunction with other clinical and laboratory findings.
Product is CE marked but not 510(k)-cleared and not available for sale in the U.S. Availability of product in each country depends on local regulatory marketing authorization status.
™ 2021 Thermo Fisher Scientific Inc. Q-linea AB is the legal manufacturer of ASTar™. ASTar™ is a trademark of Q-linea AB. All other trademarks are the property of Thermo Fisher Scientific and its subsidiaries. The ASTar system is distributed globally by Thermo Fisher Scientific.