Search
Protect large reverse osmosis systems from undissolved substances in water by using these large RO system filter insets.
| Catalog Number | Description | Pore Size (Metric) |
|---|---|---|
| 50155512 | Absolute Filter (10 in.) | 0.5 μm |
| 06.5405 | Big Blue Prefilter (20 in.) | 0.5 μm |
| 06.5406 | Big Blue Prefilter (20 in.) | 1 μm |
Protect large reverse osmosis systems from undissolved substances in water by using Thermo Scientific™ large reverse osmosis filter insets.
Big Blue filters
Ordering Information:
This product is not available in all regions