Finntip™ Universal Pipette Tips
Thermo Scientific™

Finntip™ Universal Pipette Tips

Ensure accurate, precise pipetting with these universal pipette tips, designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.

Have Questions?
Catalog number 9400220
Price (EUR)
1.088,00
Case of 20000
Price (EUR)
1.088,00
Case of 20000

Ensure accurate, precise pipetting with Thermo Scientific™ Finntip™ Universal Pipette Tips, manufactured using high quality raw materials and the latest molding techniques. Finntips are designed using our knowledge of liquid handling. Our modern automated production facilities, and our expertise, enable us to produce uniformly smooth, hydrophobic surfaces on the inside and outside of our tips to prevent liquid retention.

Features:

  • Smooth, uniform surface prevents liquid retention
  • Volume range from 0.2μL to 10mL
  • Available sterile or nonsterile
  • Packaging options include racks, bags, boxes, bulk and refill packs
  • Color-coded racks to match the compatible Finnpipette
  • Color-coding only applies to rack and refill packaging

Note:

Manufactured according to ISO 9001, ISO 14001 and ISO 13485 (CE/IVD certified).


Limited warranty period:
Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warrantied from the date we ship the product and for ninety (90) days thereafter.

Fulfills IVD Directive 98/79/EC requirements\, valid only in Europe.
Fulfills IVD Directive 98/79/EC requirements\, valid only in Europe.
Specifications
Certifications/ComplianceISO 9001, ISO 14001, ISO 13485
DescriptionFinntip 250 Universal, Non-Filtered, Non-sterile, Bulk
Length (English)2 in.
Length (Metric)5.2cm
ModelFinntip 250 Universal
Packaging20,000 tips
ColorYellow
FilteredNon-filtered
For Use With (Equipment)Thermo Scientific™ Finnpipette™ 250
FormatBulk
Product LineFinntip
SterilityNon-sterile
Tip StyleUniversal Fit
Volume (Metric)250 μL
Unit SizeCase of 20000