Finntip™ Specific Pipette Tips
Thermo Scientific™

Finntip™ Specific Pipette Tips

Ensure accurate, precise pipetting. Finntip™ Specific Pipette Tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.

Have Questions?
Catalog number 9401240
Price (EUR)
1.088,00
Case of 20000
Price (EUR)
1.088,00
Case of 20000
Ensure accurate, precise pipetting. These tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
  • Smooth, uniform surface prevents liquid retention
  • Volume range from 0.2μL to 10mL
  • Available sterile or nonsterile
  • Packaging options include racks, bags, boxes, bulk, and refill packs
  • Color-coded racks to match the compatible Finnpipette
  • Color-coding only applies to rack and refill packaging

Note:

Manufactured according to ISO 9001, ISO 14001 and ISO 13485 (CE/IVD certified)

Fulfills IVD Directive 98/79/EC requirements\, valid only in Europe.
Fulfills IVD Directive 98/79/EC requirements\, valid only in Europe.
Specifications
Certifications/ComplianceISO 9001, ISO 14001, ISO 13485
DescriptionFinntip 300, Non-Filtered, Non-sterile, Bulk
Length (English)2 in.
Length (Metric)5.2cm
ModelFinntip 300
ColorOrange
FilteredNon-filtered
For Use With (Equipment)Thermo Scientific™ Finnpipette™ 300
FormatBulk
Product LineFinntip
SterilityNon-sterile
Volume (Metric)300 μL
Unit SizeCase of 20000