1,2,4-Trichlorobenzene, 99%
1,2,4-Trichlorobenzene, 99%
1,2,4-Trichlorobenzene, 99%
Thermo Scientific Chemicals

1,2,4-Trichlorobenzene, 99%

CAS: 120-82-1 | C6H3Cl3 | 181.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
4 x 2.5 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A10130.36
also known as A10130-36
Price (EUR)
35,40
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
35,40
Each
Chemical Identifiers
CAS120-82-1
IUPAC Name1,2,4-trichlorobenzene
Molecular FormulaC6H3Cl3
InChI KeyPBKONEOXTCPAFI-UHFFFAOYSA-N
SMILESClC1=CC=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Refractive Index1.5700-1.5730 @ 20?C
CommentMay contain trace insoluble matter from the manufacturing process
Assay (GC)≥98.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
1,2,4-Trichlorobenzene is used as a dielectric and heat transfer fluid in transformers. It acts as an intermediate, degreaser, wood preservative and solvent for dye. It is a high-temperature solvent used in gel permeation chromatography, especially for polyethylene and polypropylene. Further, it is used as a lubricant and as a synthetic transformer oil.

Solubility
Miscible with ether, benzene, CS{2}.Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Bujak, M.; Podsiadło, M.; Katrusiak, A. Relations between compression and thermal contraction in 1,2,4-trichlorobenzene and melting of trichlorobenzene isomers. CrystEngComm 2015, 17 (18), 3446-3451.
  2. Dong, W. H.; Zhang, P.; Lin, X. Y.; Zhang, Y.; Tabouré, A. Natural attenuation of 1,2,4-trichlorobenzene in shallow aquifer at the Luhuagang's landfill site, Kaifeng, China. Sci. Total Environ. 2015, 505, 216-222.
  3. Su, G.; Huang, L.; Liu, S.; Lu, H.; Yang, F.; Zheng, M. The combined disposal of 1,2 4-trichlorobenzene and nitrogen oxides using the synthesized Ce0.2TiAlαOx microanomaterial. Catal. Sci. Technol. 2015, 5 (2), 1041-1051.