n-Butylidenephthalide, (E)+(Z), 95%
n-Butylidenephthalide, (E)+(Z), 95%
n-Butylidenephthalide, (E)+(Z), 95%
Thermo Scientific Chemicals

n-Butylidenephthalide, (E)+(Z), 95%

CAS: 551-08-6 | C12H12O2 | 188.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A10353.09
also known as A10353-09
Price (EUR)
38,20
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
38,20
Each
Chemical Identifiers
CAS551-08-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, yellow
FormLiquid
Assay (GC)≥94.0% (sum of isomers)
Identification (FTIR)Conforms
Refractive Index1.5740-1.5800 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only