4-Nitrophthalic acid, 97%
4-Nitrophthalic acid, 97%
4-Nitrophthalic acid, 97%
4-Nitrophthalic acid, 97%
Thermo Scientific Chemicals

4-Nitrophthalic acid, 97%

CAS: 610-27-5 | C8H5NO6 | 211.129 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
50 g
Catalog number A10368.14
also known as A10368-14
Price (EUR)
69,30
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
69,30
Each
Add to cart
Chemical Identifiers
CAS610-27-5
IUPAC Name4-nitrobenzene-1,2-dicarboxylic acid
Molecular FormulaC8H5NO6
InChI KeySLBQXWXKPNIVSQ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C=C1C(O)=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow or pale cream
FormPowder or granules
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (HPLC)≥96.0%
Melting Point (clear melt)158.5-164.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only