Molecular sieves, 13X, powder
Thermo Scientific Chemicals

Molecular sieves, 13X, powder

CAS: 63231-69-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A10378.0B
also known as A10378-0B
Price (EUR)
81,65
Online exclusive
90,10
Save 8,45 (9%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (EUR)
81,65
Online exclusive
90,10
Save 8,45 (9%)
Each
Chemical Identifiers
CAS63231-69-6
IUPAC NameMolecular sieves 13X
MDL NumberMFCD00131613
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
FormPowder
Capacity(Carbon dioxide, CO₂, adsorption): ≥18.0%
CommentNominal pore size: 10 Angstroms
CommentType X crystal sodium aluminosilicate.
View more
They are used as catalyze carrier, co adsorption of CO2 and H2O and H2O, H2O and H2S, as desiccant for medical and air compressor systems. They are normally used in gas phase and liquid phase applications. It finds its a major applications in commercial gas drying, air plant feed purification (simultaneous H2O and CO2 removal) and liquid hydrocarbon/natural gas sweetening (H2S and mercaptan removal).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only