2-Fluoro-m-xylene, 98%
2-Fluoro-m-xylene, 98%
2-Fluoro-m-xylene, 98%
Thermo Scientific Chemicals

2-Fluoro-m-xylene, 98%

CAS: 443-88-9 | C8H9F | 124.158 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A10424.06
also known as A10424-06
Price (EUR)
59,10
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
59,10
Each
Chemical Identifiers
CAS443-88-9
IUPAC Name2-fluoro-1,3-dimethylbenzene
Molecular FormulaC8H9F
InChI KeyJTAUTNBVFDTYTI-UHFFFAOYSA-N
SMILESCC1=CC=CC(C)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormLiquid
Appearance (Color)Clear colorless
Refractive Index1.4780-1.4820 @ 20?C
It is used in the spectroscopic observation of jet-cooled 2-fluoro-m-xylyl radical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the spectroscopic observation of jet-cooled 2-fluoro-m-xylyl radical.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges and keep away from ignition sources.
RUO – Research Use Only

General References:

  1. Sascha S. Zhu Dipl.-Chem, et al. Conformational Flexibility of Tetralactam Macrocycles and Their Intermolecular Hydrogen-Bonding Patterns in the Solid State.Eur. J.,2009,15(20), 5040-5046.
  2. Ted Schaefer and Reino Laatikainen. Concerning the mechanism of in toluene and its derivatives.Can. J. Chem.,1983,61(12), 2785-2789.