Diethylammonium diethyldithiocarbamate, 98%
Diethylammonium diethyldithiocarbamate, 98%
Diethylammonium diethyldithiocarbamate, 98%
Thermo Scientific Chemicals

Diethylammonium diethyldithiocarbamate, 98%

CAS: 1518-58-7 | C9H22N2S2
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A10458.14
also known as A10458-14
Price (EUR)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1518-58-7
IUPAC Namediethyl[sulfanyl(carbonothioyl)]amine; diethylamine
Molecular FormulaC9H22N2S2
InChI KeyNBGTWXBPCIHUQD-UHFFFAOYSA-N
SMILESCCNCC.CCN(CC)C(S)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤1%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)79.0-85.0?C
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Diethylammonium diethyldithiocarbamate is used in the synthesis of Dithiocarbamate. It serve as hosts for the specific binding of various metal cations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylammonium diethyldithiocarbamate is used in the synthesis of Dithiocarbamate. It serve as hosts for the specific binding of various metal cations.

Solubility
Soluble in Chloroform, Methanol.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Incompatible with strong acids, Strong bases.
RUO – Research Use Only
PF Wyatt. Diethylammonium diethyldithiocarbamate for the separation and determination of small amounts of metals. Part I. The successive determination of small amounts of copper, manganese and iron in organic compounds. Analyst.1953, 78, 656-661.