Methyl 3-iodo-4-methoxybenzoate, 98%
Methyl 3-iodo-4-methoxybenzoate, 98%
Methyl 3-iodo-4-methoxybenzoate, 98%
Thermo Scientific Chemicals

Methyl 3-iodo-4-methoxybenzoate, 98%

CAS: 35387-93-0 | C9H9IO3 | 292.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A10549.14
also known as A10549-14
Price (EUR)
169,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
169,00
Each
Chemical Identifiers
CAS35387-93-0
IUPAC Namemethyl 3-iodo-4-methoxybenzoate
Molecular FormulaC9H9IO3
InChI KeyGHNGBFHLUOJHKP-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(OC)C(I)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)90.0-97.0?C
Appearance (Color)White
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only