3-Nitrobenzoyl chloride, 98%
3-Nitrobenzoyl chloride, 98%
3-Nitrobenzoyl chloride, 98%
3-Nitrobenzoyl chloride, 98%
Thermo Scientific Chemicals

3-Nitrobenzoyl chloride, 98%

CAS: 121-90-4 | C7H4ClNO3 | 185.563 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A10579.18
also known as A10579-18
Price (EUR)
39,10
Online Exclusive
46,00
Save 6,90 (15%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (EUR)
39,10
Online Exclusive
46,00
Save 6,90 (15%)
Each
Add to cart
Chemical Identifiers
CAS121-90-4
IUPAC Name3-nitrobenzoyl chloride
Molecular FormulaC7H4ClNO3
InChI KeyNXTNASSYJUXJDV-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC(=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormFused solid
Melting Point (clear melt)29.0-36.0?C
Identification (FTIR)Conforms
Appearance (Color)White to pale yellow to pale brown
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only