3-Fluoro-o-xylene, 99%
3-Fluoro-o-xylene, 99%
3-Fluoro-o-xylene, 99%
3-Fluoro-o-xylene, 99%
Thermo Scientific Chemicals

3-Fluoro-o-xylene, 99%

CAS: 443-82-3 | C8H9F | 124.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A10637.22
also known as A10637-22
Price (EUR)
295,00
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
295,00
Each
Add to cart
Chemical Identifiers
CAS443-82-3
IUPAC Name1-fluoro-2,3-dimethylbenzene
Molecular FormulaC8H9F
InChI KeyAWLDSXJCQWTJPC-UHFFFAOYSA-N
SMILESCC1=CC=CC(F)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
FormLiquid
Refractive Index1.4835-1.4865 @ 20?C
Appearance (Color)Clear colorless
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Stable under normal conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Young Wook Yoon; Sang Youl Chae; Sang Kuk Lee. Spectroscopic identification of isomeric jet-cooled benzyl-type radicals formed from 3-fluoro-o-xylene by corona discharge. Chemical Physics Letters. 2013, 584, 37-42