Pyridine-4-carboxaldehyde, 97%
Pyridine-4-carboxaldehyde, 97%
Pyridine-4-carboxaldehyde, 97%
Pyridine-4-carboxaldehyde, 97%
Thermo Scientific Chemicals

Pyridine-4-carboxaldehyde, 97%

CAS: 872-85-5 | C6H5NO | 107.112 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A10656.14
also known as A10656-14
Price (EUR)
35,02
キャンペーン価格
41,20
Save 6,18 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
35,02
キャンペーン価格
41,20
Save 6,18 (15%)
Each
Add to cart
Chemical Identifiers
CAS872-85-5
IUPAC Namepyridine-4-carbaldehyde
Molecular FormulaC6H5NO
InChI KeyBGUWFUQJCDRPTL-UHFFFAOYSA-N
SMILESO=CC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to yellow to brown
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5415-1.5465 @ 20?C
Pyridine-4-carboxaldehyde is an heterocyclic building block used for the synthesis of various pharmaceutical compounds, such as new 1,4-dihydropyridin-4-yl-phenoxyacetohydrazones, having anticonvulsant and anti-inflammatory properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine-4-carboxaldehyde is an heterocyclic building block used for the synthesis of various pharmaceutical compounds, such as new 1,4-dihydropyridin-4-yl-phenoxyacetohydrazones, having anticonvulsant and anti-inflammatory properties.

Solubility
Soluble in water (20 mg/ml at 20°C), ethanol and acetone.

Notes
It is air and light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Nand K.Singh; Rajesh Tripathi. Synthesis and structural characterization of 3d-metal complexes of pyridine-4-carboxaldehyde thionicotinoyl hydrazone. Transition Metal Chemistry. 1988, 13, (5),346-350
  2. A.Martin; B.Lucke; H.J.Niclas; A.Forster. Vapor-phase oxidation of 4-picoline to pyridine-4-carboxaldehyde on a vanadium phosphate catalysts. Reaction Kinetics and Catalysis Letters. 1991, 43, (2),583-588
  3. In the presence of DBU, converts amines to carbonyl compounds, e.g. benzylamines to benzaldehydes: Synthesis, 756 (1982):
  4. The reaction is also useful for the conversion of ɑ-amino acids to ɑ-keto acids.