m-Cresol, 99%
m-Cresol, 99%
m-Cresol, 99%
Thermo Scientific Chemicals

m-Cresol, 99%

CAS: 108-39-4 | C7H8O | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
800 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A10733.30
also known as A10733-30
Price (EUR)
32,90
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
32,90
Each
Chemical Identifiers
CAS108-39-4
IUPAC Name3-methylphenol
Molecular FormulaC7H8O
InChI KeyRLSSMJSEOOYNOY-UHFFFAOYSA-N
SMILESCC1=CC=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥98.5%
CommentMaterial Sourced in UK and US
Appearance (Color)Clear colorless to yellow
Refractive Index1.5390-1.5420 @ 20?C (UK Sourced Material)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
m-Cresol is employed in the preparation of synthetic resins like phenol-formaldehyde resins, disinfectants, and photographic developers. It is used as industrial solvent for dissolving polymers such as polyaniline. It acts as a precursor to vitamin E and antiseptic amylmetacresol. It is used as insuline preservatives.

Solubility
Miscible with water, diethylether, chloroform and ethanol.

Notes
Hygroscopic. Air and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases, strong acids, aluminum and aluminum alloys.
RUO – Research Use Only

General References:

  1. Liu, P.; He, S.; Wei, H.; Wang, J.; Sun, C. Characterization of alpha-Fe2O3/gamma-Al2O3 Catalysts for Catalytic Wet Peroxide Oxidation of m-Cresol. Ind. Eng. Chem. Res. 2015, 54 (1), 130-136.
  2. Tan, Q.; Wang, G.; Nie, L.; Dinse, A.; Buda, C.; Shabaker, J.; Resasco, D. E. Different Product Distributions and Mechanistic Aspects of the Hydrodeoxygenation of m-Cresol over Platinum and Ruthenium Catalysts. ACS Catal. 2015, 5 (11), 6271-6283.