Ethyl 2-bromopropionate, 98+%
Ethyl 2-bromopropionate, 98+%
Ethyl 2-bromopropionate, 98+%
Thermo Scientific Chemicals

Ethyl 2-bromopropionate, 98+%

CAS: 535-11-5 | C5H9BrO2 | 181.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
500 g
Catalog number A10820.18
also known as A10820-18
Price (EUR)
27,00
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
27,00
Each
Chemical Identifiers
CAS535-11-5
IUPAC Nameethyl 2-bromopropanoate
Molecular FormulaC5H9BrO2
InChI KeyARFLASKVLJTEJD-UHFFFAOYNA-N
SMILESCCOC(=O)C(C)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
Assay (GC)≥98.0%
Refractive Index1.4440-1.4480 @ 20?C
FormLiquid
It’s an important organic intermediate can be used in agrochemical, pharmaceutical synthesis and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It’s an important organic intermediate can be used in agrochemical, pharmaceutical synthesis and dyestuff field.

Solubility
Immiscible in water. Soluble in alcohol and ether.

Notes
Store in dark and away from oxidizing agents and acids.
RUO – Research Use Only

General References:

  1. T.S. Jagodziñski.; A. Weso³owska.; J.G. Soœnicki. Mechanism of levulinic acid formation. Tetrahedron Letters. 2000, 74 1101-1114.
  2. Xiao-Song Wang.; Ning Luo.; Sheng-Kang Ying. Controlled/living polymerization of MMA promoted by heterogeneous initiation system (EPN-X-CuX-bpy). Journal of Polymer Science Part A: Polymer Chemistry. 1999, 37 (9), 1255-1263.