2,5-Dimethylthiophene, 98+%
2,5-Dimethylthiophene, 98+%
2,5-Dimethylthiophene, 98+%
Thermo Scientific Chemicals

2,5-Dimethylthiophene, 98+%

CAS: 638-02-8 | C6H8S | 112.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Catalog number A10869.09
also known as A10869-09
Price (EUR)
40,65
Online exclusive
44,90
Save 4,25 (9%)
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
40,65
Online exclusive
44,90
Save 4,25 (9%)
Each
Chemical Identifiers
CAS638-02-8
IUPAC Name2,5-dimethylthiophene
Molecular FormulaC6H8S
InChI KeyGWQOOADXMVQEFT-UHFFFAOYSA-N
SMILESCC1=CC=C(C)S1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow to orange
Refractive Index1.5105-1.5145 @ 20°C
Identification (FTIR)Conforms
Assay (GC)≥98.0%
2,5-Dimethylthiophene is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethylthiophene is used as an intermediate in organic synthesis.

Solubility
Insoluble in water. Soluble in alcohol, ether and benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Deborah R. Huntley.; David R. Mullins.; Michael P. Wingeier. Desulfurization of Thiophenic Compounds by Ni(111): Adsorption and Reactions of Thiophene, 3-Methylthiophene, and 2,5-Dimethylthiophene. J. Phys. Chem. 1996, 100, (50), 19620-19627.
  2. Jiabi Chen.; Robert J. Angelici. 2,5-Dimethylthiophene coordination to three metal centers in (.eta.4,S-.mu.3-2,5-Me2T)(IrCp*)[Mo(CO)2Cp]2. Organometallics. 1990, 9, (3), 879-880.