Ethyl L-lactate, 99%
Ethyl L-lactate, 99%
Ethyl L-lactate, 99%
Thermo Scientific Chemicals

Ethyl L-lactate, 99%

CAS: 687-47-8 | C5H10O3 | 118.132 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 g
500 g
2500 g
Catalog number A10900.0C
also known as A10900-0C
Price (EUR)
384,00
Each
Quantity:
10,000 g
Request bulk or custom format
Price (EUR)
384,00
Each
Chemical Identifiers
CAS687-47-8
IUPAC Nameethyl (2S)-2-hydroxypropanoate
Molecular FormulaC5H10O3
InChI KeyLZCLXQDLBQLTDK-BYPYZUCNSA-N
SMILESCCOC(=O)[C@H](C)O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4115-1.4145 @ 20°C
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Identification (FTIR)Conforms
View more
Ethyl L-lactate is used in the preparation of food additives, aryl aldimines, fragrances and in pharmaceutical preparations. It can be considered as a green solvent in chemical industries due to its biodegradable nature. As a solvent, it used in the production of nitrocellulose, cellulose acetate and cellulose ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl L-lactate is used in the preparation of food additives, aryl aldimines, fragrances and in pharmaceutical preparations. It can be considered as a green solvent in chemical industries due to its biodegradable nature. As a solvent, it used in the production of nitrocellulose, cellulose acetate and cellulose ethers.

Solubility
Miscible with water and chloroform.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Ethyl (or isobutyl) L-lactate adds to aryl methyl ketenes to give esters of 2-arylpropionic acids in which the chirality is transferred to the new chiral center to a surprising degree: J. Am. Chem. Soc., 111, 7650 (1989):
  2. Silylation followed by reaction with MeLi gives a useful chiral synthon. For preparation and discussion of reactivity, see: Org. Synth. Coll., 9, 139 (1998):
  3. For reaction with Diphenyl phosphonic azide, A12124, in the prepartaion, with inversion, of ethyl (R)-2-azidopropionate, see: Org. Synth., 75, 31 (1997).
  4. Upare, P. P.; Lee, M.; Hwang, D. W.; Hwang, Y. K.; Chang, J. S. New heterogeneous Pb oxide catalysts for lactide production from an azeotropic mixture of ethyl lactate and water. Catal. Commun. 2014, 56, 179-183.
  5. Williams, J. D.; Torhan, M. C.; Neelagiri, V. R.; Brown, C.; Bowlin, N. O.; Di, M.; Moir, D. T. Synthesis and structure-activity relationships of novel phenoxyacetamide inhibitors of the Pseudomonas aeruginosa type III secretion system (T3SS). Bioorg. Med. Chem. 2015, 23 (5), 1027-1043.