4-tert-Butylaniline, 98+%
4-tert-Butylaniline, 98+%
4-tert-Butylaniline, 98+%
Thermo Scientific Chemicals

4-tert-Butylaniline, 98+%

CAS: 769-92-6 | C10H15N | 149.237 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A10925.06
also known as A10925-06
Price (EUR)
60,80
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
60,80
Each
Chemical Identifiers
CAS769-92-6
IUPAC Name4-tert-butylaniline
Molecular FormulaC10H15N
InChI KeyWRDWWAVNELMWAM-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.5365-1.5395 @ 20°C
Appearance (Color)Clear colorless to pale yellow to yellow to orange or pale pink
Identification (FTIR)Conforms
4-tert-Butylaniline reacts with formic acid to produce formic acid-(4-tert-butyl-anilide). This reaction could happen in the presence of the solvent of toluene in the condition of heating. 4-tert-Butylaniline is used in the synthesis of 4-tert-Butyl-4?,4?-dinitrotriphenylamine, new triphenylamine-containing diamine monomer and 2-oxopyrimido[4,5-d]pyrimidin-5(6H)-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylaniline reacts with formic acid to produce formic acid-(4-tert-butyl-anilide). This reaction could happen in the presence of the solvent of toluene in the condition of heating. 4-tert-Butylaniline is used in the synthesis of 4-tert-Butyl-4′,4″-dinitrotriphenylamine, new triphenylamine-containing diamine monomer and 2-oxopyrimido[4,5-d]pyrimidin-5(6H)-one.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to light. Incompatible with strong acids, acid chlorides, acid anhydrides and chloroformates.
RUO – Research Use Only

General References:

  1. J. Doucet.; A. M. Levelut.; M. Lambert. Polymorphism of the Mesomorphic Compound Terephthal-bis-Butylaniline (TBBA). Phys. Rev. Lett. 1974, 32, (6), 301.
  2. Jean Yves Bergeron.; Le H. Dao. Electrical and physical properties of new electrically conducting quasi-composites. Poly(aniline-co-N-butylaniline) copolymers. Macromolecules. 1992, 25, (13), 3332-3337