S-Methylisothiouronium sulfate, 98+%
S-Methylisothiouronium sulfate, 98+%
S-Methylisothiouronium sulfate, 98+%
Thermo Scientific Chemicals

S-Methylisothiouronium sulfate, 98+%

CAS: 867-44-7 | C4H14N4O4S3 | 278.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
500 g
2500 g
Catalog number A11044.22
also known as A11044-22
Price (EUR)
27,10
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
27,10
Each
Chemical Identifiers
CAS867-44-7
IUPAC Namebis((methylsulfanyl)methanimidamide); sulfuric acid
Molecular FormulaC4H14N4O4S3
InChI KeyBZZXQZOBAUXLHZ-UHFFFAOYSA-N
SMILESCSC(N)=N.CSC(N)=N.OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
FormCrystals or powder or crystalline powder or fused solid
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only