Antipyrine, 98%
Antipyrine, 98%
Antipyrine, 98%
Thermo Scientific Chemicals

Antipyrine, 98%

CAS: 60-80-0 | C11H12N2O | 188.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A11089.22
also known as A11089-22
Price (EUR)
36,40
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
36,40
Each
Chemical Identifiers
CAS60-80-0
IUPAC Name1,5-dimethyl-2-phenyl-2,3-dihydro-1H-pyrazol-3-one
Molecular FormulaC11H12N2O
InChI KeyVEQOALNAAJBPNY-UHFFFAOYSA-N
SMILESCN1N(C(=O)C=C1C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White
FormCrystals or powder or crystalline powder
Melting Point (clear melt)108.0-114.0?C
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only