Ammonium iron(III) citrate, brown
Ammonium iron(III) citrate, brown
Ammonium iron(III) citrate, brown
Ammonium iron(III) citrate, brown
Thermo Scientific Chemicals

Ammonium iron(III) citrate, brown

CAS: 1185-57-5 | C6H11FeNO7+3 | 264.999 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A11199.30
also known as A11199-30
Price (EUR)
23,72
Online Exclusive
27,90
Save 4,18 (15%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (EUR)
23,72
Online Exclusive
27,90
Save 4,18 (15%)
Each
Add to cart
Chemical Identifiers
CAS1185-57-5
IUPAC Nameiron(3+) 2-hydroxypropane-1,2,3-tricarboxylate amine
Molecular FormulaC6H8FeNO7
InChI KeyFRHBOQMZUOWXQL-UHFFFAOYSA-K
SMILESN.[Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown-yellow to brown
FormCrystals or crystalline powder or powder
Assay (unspecified)Fe: 16.5-18.5%
Ammonium iron(III) citrate, brown is used in medicine, in making blueprints, and as a feed additive. It is also used as an iron supplement and food enhancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium iron(III) citrate, brown is used in medicine, in making blueprints, and as a feed additive. It is also used as an iron supplement and food enhancer.

Solubility
Soluble in water.1200 g/L (20°C)

Notes
Hygroscopic. Desiccate at room temperature. Store in the dark. Store under inert gas.
RUO – Research Use Only

General References:

  1. Malcolm P. N. Brown J. J. Hahn, P. F. Stillman, A. E. Li, K. C.P., Kawamura Y. Tanaka, T. Noel J. K. Molony B. A. Johnson M. F. and Hildebolt C. F. The clinical value of ferric ammonium citrate: A positive oral contrast agent for T1-weighted MR imaging of the upper abdomen. J. Magn. Reson. Imaging. 2000, 702-707.
  2. D Richardson and E.baker. Two mechanisms of iron uptake from transferrin by melanoma cells. The effect of desferrioxamine and ferric ammonium citrate. Journal of biological chemistry. 1992, 267, 13972-13979.