3,4-Dichlorophenylacetonitrile, 98%
3,4-Dichlorophenylacetonitrile, 98%
3,4-Dichlorophenylacetonitrile, 98%
Thermo Scientific Chemicals

3,4-Dichlorophenylacetonitrile, 98%

CAS: 3218-49-3 | C8H5Cl2N | 186.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A11382.22
also known as A11382-22
Price (EUR)
118,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
118,00
Each
Chemical Identifiers
CAS3218-49-3
IUPAC Name2-(3,4-dichlorophenyl)acetonitrile
Molecular FormulaC8H5Cl2N
InChI KeyQWZNCAFWRZZJMA-UHFFFAOYSA-N
SMILESClC1=CC=C(CC#N)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White to cream
FormCrystalline or fused solid, lumps or powder
Melting Point (clear melt)38-43?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only