4,4'-Dimethoxybenzophenone, 98+%
4,4'-Dimethoxybenzophenone, 98+%
4,4'-Dimethoxybenzophenone, 98+%
4,4'-Dimethoxybenzophenone, 98+%
Thermo Scientific Chemicals

4,4'-Dimethoxybenzophenone, 98+%

CAS: 90-96-0 | C15H14O3 | 242.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A11466.14
also known as A11466-14
Price (EUR)
49,70
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
49,70
Each
Add to cart
Chemical Identifiers
CAS90-96-0
IUPAC Namebis(4-methoxyphenyl)methanone
Molecular FormulaC15H14O3
InChI KeyRFVHVYKVRGKLNK-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C(=O)C1=CC=C(OC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Appearance (Color)White
FormCrystals or powder or crystalline powder
Melting Point (clear melt)141.0-148.0?C
4,4'-Dimethoxybenzophenone is used in the organic synthesis. It also acts as a pharmaceutical intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dimethoxybenzophenone is used in the organic synthesis. It also acts as a pharmaceutical intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
It is insoluble in water, soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents, strong reducing agents.
RUO – Research Use Only

General References:

  1. Babatunde B. Adeleke.; Kuang S. Chen.; Jeffrey K.S. Wan. A comparative esr study of the reactions of group ivb organometallic radicals with 4,4'-dimethoxybenzophenone, 4,4'-dimethoxythiobenzophenone, and 3-ethyl-2-thioxo-4-oxazolidione. J. Organometallic. Chem. 1981, 208, (3), 317-326.
  2. Oksana Shvydkiv.; Kieran Nolan .; Michael Oelgemöller. Microphotochemistry: 4,4'-Dimethoxybenzophenone mediated photodecarboxylation reactions involving phthalimides. Beilstein J. Org. Chem. 2011, , (7), 1055-1063.