Hypoxanthine, 99%
Hypoxanthine, 99%
Hypoxanthine, 99%
Thermo Scientific Chemicals

Hypoxanthine, 99%

CAS: 68-94-0 | C5H4N4O | 136.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11481.14
also known as A11481-14
Price (EUR)
81,80
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
81,80
Each
Chemical Identifiers
CAS68-94-0
IUPAC Name6,7-dihydro-3H-purin-6-one
Molecular FormulaC5H4N4O
InChI KeyFDGQSTZJBFJUBT-UHFFFAOYSA-N
SMILESO=C1N=CNC2=C1NC=N2
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.5%
FormPowder or lumps
Appearance (Color)White to pale cream
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Hypoxanthine, is used as a marker for energy perturbation in hypoxia/ ischemia. It has been also used in DNA studies to investigate the destabilizing effect it has on DNA duplexes containing hypoxanthine as a base, in a gas phase versus a liquid phase. Hypoxanthine has also been used in studies along with 8-oxoguanine nucleotides to investigate their interaction with human DNA pol A (DNA polymerase alpha) and DNA polymerase I from Bacillus stearothermophilus. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hypoxanthine, is used as a marker for energy perturbation in hypoxia/ ischemia. It has been also used in DNA studies to investigate the destabilizing effect it has on DNA duplexes containing hypoxanthine as a base, in a gas phase versus a liquid phase. Hypoxanthine has also been used in studies along with 8-oxoguanine nucleotides to investigate their interaction with human DNA pol å (DNA polymerase alpha) and DNA polymerase I from Bacillus stearothermophilus. It is also used as a pharmaceutical intermediate.

Solubility
It does not mix appreciably with water (1:1400). Small octahedra crystals from water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents. Stable under recommended storage conditions. Sterilize by filtration through a 0.22 µm filter.
RUO – Research Use Only

General References:

  1. Patrick A. Limbach.; Pamela F. Crain .; James A. McCloskey. the modified nucleosides of RNA. Oxford Journals. 1994, 22, (12), 2183-2196.
  2. James S. Chickos .; William E. Acree Jr. Enthalpies of Sublimation of Organic and Organometallic Compounds. J. Phys. Chem.. 2002, 31, 537.