1-Chlorobutane, 99+%
1-Chlorobutane, 99+%
1-Chlorobutane, 99+%
Thermo Scientific™

1-Chlorobutane, 99+%

CAS: 109-69-3 | C4H9Cl | 92.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A11494.AP
Price (EUR)
28,50
Each
Quantity:
500 mL
Request bulk or custom format
Price (EUR)
28,50
Each
Chemical Identifiers
CAS109-69-3
IUPAC Name1-chlorobutane
Molecular FormulaC4H9Cl
InChI KeyVFWCMGCRMGJXDK-UHFFFAOYSA-N
SMILESCCCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4010-1.4035 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥99.0%
FormLiquid
Identification (FTIR)Conforms
1-Chlorobutane is used as intermediates and solvents. It is used for HPLC, spectrophotometry and environmental testing. It is used as an alkylating reagent in organic and organometallic compound synthesis. It is also employed as an anthelmintic in veterinary medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chlorobutane is used as intermediates and solvents. It is used for HPLC, spectrophotometry and environmental testing. It is used as an alkylating reagent in organic and organometallic compound synthesis. It is also employed as an anthelmintic in veterinary medicine.

Solubility
Miscible with alcohol and ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. The Grignard reagent has been recommended as superior to EtMgBr for the preparation of ethynyl magnesium halides from acetylene. For details, see: Org. Synth. Coll., 8, 606 (1993); for generation and use of the Grignard in the Mn-Cu catalyzed 1,4-addition to an enone, see: Org. Synth. Coll., 9, 328 (1998).
  2. Mututuvari, T. M.; Tran, C. D. Volumetric Study for the Binary Nitromethane with Chloroalkane Mixtures at Temperatures in the Range (298.15 to 318.15) K. J. Hazard. Mater. 2014, 264, 449-459.
  3. Atanase, L. I.; Bistac, S.; Riess, G. Effect of poly(vinyl alcohol-co-vinyl acetate) copolymer blockiness on the dynamic interfacial tension and dilational viscoelasticity of polymer-anionic surfactant complex at the water-1-chlorobutane interface. Soft matter 2015, 11 (13), 2665-2672.