4-Bromocinnamic acid, predominantly trans, 97+%
4-Bromocinnamic acid, predominantly trans, 97+%
4-Bromocinnamic acid, predominantly trans, 97+%
Thermo Scientific Chemicals

4-Bromocinnamic acid, predominantly trans, 97+%

CAS: 1200-07-3 | C9H7BrO2 | 227.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11503.14
also known as A11503-14
Price (EUR)
113,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
113,00
Each
Chemical Identifiers
CAS1200-07-3
IUPAC Name(2E)-3-(4-bromophenyl)prop-2-enoic acid
Molecular FormulaC9H7BrO2
InChI KeyCPDDDTNAMBSPRN-ZZXKWVIFSA-N
SMILESOC(=O)C=CC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)259.5-265.5°C
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥97.0%
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.0 to ≤103.0%
4-Bromocinnamic acid has been used in the preparation of: (E)-β-bromo-4-bromostyrene, 2-amino-7-(piperidin-4-yl)isoquinoline, brominated dansyl derivative (4-bromophenyl)-4-(5-(dimethylamino)naphthalene-1-sulfonamido)butanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromocinnamic acid has been used in the preparation of: (E)-β-bromo-4-bromostyrene, 2-amino-7-(piperidin-4-yl)isoquinoline, brominated dansyl derivative (4-bromophenyl)-4-(5-(dimethylamino)naphthalene-1-sulfonamido)butanoic acid.

Solubility
Soluble in alcohol, ethyl acetate, DMSO. Insoluble in water.

Notes
Store at -20°C. Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
RUO – Research Use Only
Jane L. R. Yates and Hazel A. Sparkes. 4-Bromo-trans-cinnamic acid: structural characterization and crystallographic investigation into the solid state [2 + 2] cycloaddition reaction and temperature induced phase transition. CrystEngComm.2013, 15 3547-3553.