Diethyl dibromomalonate, 96%
Diethyl dibromomalonate, 96%
Diethyl dibromomalonate, 96%
Thermo Scientific Chemicals

Diethyl dibromomalonate, 96%

CAS: 631-22-1 | C7H10Br2O4 | 317.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11508.06
also known as A11508-06
Price (EUR)
22,60
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
22,60
Each
Chemical Identifiers
CAS631-22-1
IUPAC Name1,3-diethyl 2,2-dibromopropanedioate
Molecular FormulaC7H10Br2O4
InChI KeyPFZYFZRUPFUEOB-UHFFFAOYSA-N
SMILESCCOC(=O)C(Br)(Br)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥95.0%
Refractive Index1.4815-1.4865 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only