Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate, 98%
Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate, 98%
Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate, 98%
Thermo Scientific Chemicals

Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate, 98%

CAS: 31037-02-2 | C7H11N3O2 | 169.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A11534.06
also known as A11534-06
Price (EUR)
41,20
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
41,20
Each
Chemical Identifiers
CAS31037-02-2
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)97.0-103.0?C
Appearance (Color)Pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only