Sodium dodecatungstophosphate hydrate
Sodium dodecatungstophosphate hydrate
Sodium dodecatungstophosphate hydrate
Thermo Scientific Chemicals

Sodium dodecatungstophosphate hydrate

CAS: 51312-42-6 | Na3O40PW12 | 2946.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A11575.36
also known as A11575-36
Price (EUR)
676,00
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
676,00
Each
Chemical Identifiers
CAS51312-42-6
IUPAC NameSodium dodecamolybdophosphate hydrate
Molecular FormulaNa3O40PW12
SMILES*
Molecular Weight (g/mol)2946.12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Elemental AnalysisTungsten (W) ≥ 68.5%
Elemental AnalysisSodium ≥ 1.25%
Solution Test100mg/ml water: clear
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only