2,4-Difluorophenylhydrazine hydrochloride, 97%
2,4-Difluorophenylhydrazine hydrochloride, 97%
2,4-Difluorophenylhydrazine hydrochloride, 97%
Thermo Scientific Chemicals

2,4-Difluorophenylhydrazine hydrochloride, 97%

CAS: 51523-79-6 | C6H6F2N2 | 144.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11625.22
also known as A11625-22
Price (EUR)
662,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
662,00
Each
Chemical Identifiers
CAS51523-79-6
IUPAC Name(2,4-difluorophenyl)hydrazine
Molecular FormulaC6H6F2N2
InChI KeyRLZUIPTYDYCNQI-UHFFFAOYSA-N
SMILESNNC1=CC=C(F)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only