Ammonium sulfate, 98+%
Ammonium sulfate, 98+%
Ammonium sulfate, 98+%
Thermo Scientific Chemicals

Ammonium sulfate, 98+%

CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
5000 g
Catalog number A11682.0I
also known as A11682-0I
Price (EUR)
114,65
Online exclusive
127,00
Save 12,35 (10%)
Each
Quantity:
5000 g
Request bulk or custom format
Price (EUR)
114,65
Online exclusive
127,00
Save 12,35 (10%)
Each
Chemical Identifiers
CAS7783-20-2
IUPAC Namesulfuric acid diamine
Molecular FormulaH8N2O4S
InChI KeyBFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILESN.N.OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Ammonium sulfate is widely utilized as a fertilizer for alkaline soils. In biochemistry it is used for purification of proteins via ammonium sulfate precipitation method. As a food additive, it serves as an acidity regulator in flours and breads. It is used in the preparation of other ammonium salts, particularly ammonium persulfate. It is applied in several applications as a flame retardant and a wood preservative. It finds use as raw materials for various chemical products such as dyes and pharmaceutical products. A saturated solution of ammonium sulfate in heavy water is employed as an external standard in sulfur-33 NMR spectroscopy.

Solubility
Soluble in water. Insoluble in alcohol and acetone.

Notes
Incompatible with various substances like highly reactive oxidizing agents and alkalis.
RUO – Research Use Only

General References:

  1. Guo, H.; Chen, W.; Sun, X.; Liu, Y. N.; Li, J.; Wang, J. Theranostic magnetoliposomes coated by carboxymethyl dextran with controlled release by low-frequency alternating magnetic field. Carbohydr. Polym. 2015, 118, 209-217.
  2. Carter, D.; Rose, L.; Awobusuyi, T.; Gauthier, M.; Tezel, F. H.; Kruczek, B. Characterization of commercial RO membranes for the concentration of ammonia converted to ammonium sulfate from anaerobic digesters. Desalination 2015, 368, 127-134.