2-(1-Piperidinyl)benzonitrile, 97%
2-(1-Piperidinyl)benzonitrile, 97%
2-(1-Piperidinyl)benzonitrile, 97%
Thermo Scientific Chemicals

2-(1-Piperidinyl)benzonitrile, 97%

CAS: 72752-52-4 | C12H14N2 | 186.258 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A11784.03
also known as A11784-03
Price (EUR)
13,60
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
13,60
Each
Chemical Identifiers
CAS72752-52-4
IUPAC Name2-(piperidin-1-yl)benzonitrile
Molecular FormulaC12H14N2
InChI KeyMEBVSLLKZSAIGK-UHFFFAOYSA-N
SMILESN#CC1=CC=CC=C1N1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)37.0-44.0°C
Assay (GC)≥96.0%
Appearance (Color)White
FormCrystals or powder or crystalline powder or chunks
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only