2-Chlorobenzyl chloride, 98+%
2-Chlorobenzyl chloride, 98+%
2-Chlorobenzyl chloride, 98+%
2-Chlorobenzyl chloride, 98+%
Thermo Scientific Chemicals

2-Chlorobenzyl chloride, 98+%

CAS: 611-19-8 | C7H6Cl2 | 161.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
2500 g
Catalog number A11900.0E
also known as A11900-0E
Price (EUR)
173,00
Each
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (EUR)
173,00
Each
Add to cart
Chemical Identifiers
CAS611-19-8
IUPAC Name1-chloro-2-(chloromethyl)benzene
Molecular FormulaC7H6Cl2
InChI KeyBASMANVIUSSIIM-UHFFFAOYSA-N
SMILESClCC1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5570-1.5610 @ 20?C
FormLiquid
Identification (FTIR)Conforms
2-Chlorobenzyl chloride was used as a sensory irritant to evaluate the characteristic modifications of the normal breathing pattern of exposed mice. It is used as an intermediate for organic chemicals, pharmaceuticals, and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzyl chloride was used as a sensory irritant to evaluate the characteristic modifications of the normal breathing pattern of exposed mice. It is used as an intermediate for organic chemicals, pharmaceuticals, and dyes.

Solubility
Not miscible in water.

Notes
Store away from strong bases and oxidizing agents. Keep away from heat. Incompatible with Alcohols, Amines, Metals.
RUO – Research Use Only

General References:

  1. Robert S. Schwab; Clare K. Marshall; William Timberlake. Win 8077 in treatment of myasthenia gravisuse of n,n'-bis (2-diethylaminoethyl) oxamide bis-2-chlorobenzyl chloride in fifty patients.JAMA. 1955, 158 (8), 625-628..