Chloroacetone was used in the synthesis of double-chain nonionic surfactants with an acid decomposition function via acid-catalyzed condensation with fatty alcohols (octyl, decyl and dodecyl). It was also used in the synthesis of meso-tetramethyl tetrakis-(4-phenoxy acetone)calix[4]pyrrole. It can also be used to make dye couplers for color photography and used in the Feist-Benary synthesis of furans.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chloroacetone was used in the synthesis of double-chain nonionic surfactants with an acid decomposition function via acid-catalyzed condensation with fatty alcohols (octyl, decyl and dodecyl). It was also used in the synthesis of meso-tetramethyl tetrakis-(4-phenoxy acetone)calix[4]pyrrole. It can also be used to make dye couplers for color photography and used in the Feist-Benary synthesis of furans.
Solubility
Soluble in water (124g/L), ether and chloroform.
Notes
Refrigerate. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Do not store together with strong basic or oxidizing materials.
RUO – Research Use Only
General References:
- Weijers CAGM, et al. Synthesis of optically pure 1, 2-epoxypropane by microbial asymmetric reduction of chloroacetone. Appl. Microbiol. Biotechnol.1992,38(3), 297-300.
- He L, et al. L-Proline amide-catalyzed direct asymmetric aldol reaction of aldehydes with chloroacetone. Tetrahedron,2006, 62(2), 590-599.