2,4-Dimethylimidazole, 97%
2,4-Dimethylimidazole, 97%
2,4-Dimethylimidazole, 97%
Thermo Scientific Chemicals

2,4-Dimethylimidazole, 97%

CAS: 930-62-1 | C5H8N2 | 96.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A11949.06
also known as A11949-06
Price (EUR)
95,40
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
95,40
Each
Chemical Identifiers
CAS930-62-1
IUPAC Name2,5-dimethyl-1H-imidazole
Molecular FormulaC5H8N2
InChI KeyLLPKQRMDOFYSGZ-UHFFFAOYSA-N
SMILESCC1=CN=C(C)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)White to pale yellow or cream
FormCrystals or powder or crystalline powder and/or chunks
Water Content (Karl Fischer Titration)≤2.0%
It is employed as a curing agent of epoxy resins. It is also widely used in the bonding, coating, capsulation of epoxy resins, or used for composite materials. It is also employed as medical raw material or used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a curing agent of epoxy resins. It is also widely used in the bonding, coating, capsulation of epoxy resins, or used for composite materials. It is also employed as medical raw material or used in organic synthesis.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
RUO – Research Use Only

General References:

  1. Yasuhiko Nozaki.; Frank R. N. Gurd.; Raymond F. Chen.; John T. Edsall. The Association of 4-Methylimidazole with the Ions of Cupric Copper and Zinc; with Some Observations on 2,4-Dimethylimidazole.J. Am. Chem. Soc.,1957,79(9), 2123-2129.
  2. Yasuhiro Ishida.; Daisuke Sasaki.; Hiroyuki Miyauchi.; Kazuhiko Saigo. Design and synthesis of novel imidazolium-based ionic liquids with a pseudo crown-ether moiety: diastereomeric interaction of a racemic ionic liquid with enantiopure europium complexes.Tetrahedron Lett.2004,45(51), 9455-9459.