(+/-)-Phenylsuccinic acid, 98%
(+/-)-Phenylsuccinic acid, 98%
(+/-)-Phenylsuccinic acid, 98%
Thermo Scientific Chemicals

(+/-)-Phenylsuccinic acid, 98%

CAS: 635-51-8 | C10H10O4 | 194.186 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A11971.22
also known as A11971-22
Price (EUR)
67,65
Online exclusive
74,90
Save 7,25 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
67,65
Online exclusive
74,90
Save 7,25 (10%)
Each
Chemical Identifiers
CAS635-51-8
IUPAC Name2-phenylbutanedioic acid
Molecular FormulaC10H10O4
InChI KeyLVFFZQQWIZURIO-UHFFFAOYNA-N
SMILESOC(=O)CC(C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point165-169°C
Appearance (Color)White to cream
FormPowder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only