Hexa-n-butylditin, 97%
Hexa-n-butylditin, 97%
Hexa-n-butylditin, 97%
Thermo Scientific Chemicals

Hexa-n-butylditin, 97%

CAS: 813-19-4 | C24H54Sn2 | 580.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A12007.30
also known as A12007-30
Price (EUR)
632,00
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
632,00
Each
Chemical Identifiers
CAS813-19-4
IUPAC Namehexabutyldistannane
Molecular FormulaC24H54Sn2
InChI KeyREDSKZBUUUQMSK-UHFFFAOYSA-N
SMILESCCCC[Sn](CCCC)(CCCC)[Sn](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index1.5105-1.5145 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
Hexa-n-butylditin is used in palladium-catalyzed tin-carbon bond formation. It finds useful application in deoxygenation and desulfurization reactions. It acts as a source of tributylstannyl radicals in synthetic chemistry. It is also used in the radioiodination of styrlpyridines. Further, it is used as a reagent to stannylate aryl halides for further Stille coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexa-n-butylditin is used in palladium-catalyzed tin-carbon bond formation. It finds useful application in deoxygenation and desulfurization reactions. It acts as a source of tributylstannyl radicals in synthetic chemistry. It is also used in the radioiodination of styrlpyridines. Further, it is used as a reagent to stannylate aryl halides for further Stille coupling.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Source of on thermolysis or photolysis of tributylstannyl radicals, avoiding the problems sometimes attributable to the hydrogen donor capability of Tri-n-butyl tin hydride, A13298 .ɑ-Iodo ketones or esters with a suitably-placed double bond undergo radical cyclizations on UV irradiation in the presence of a catalytic amount of the distannane: Tetrahedron Lett., 28, 2477 (1987); J. Org. Chem., 54, 1826, 3140 (1989). Compare analogous reductive cyclizations using the tin hydride. Improved conditions have been developed for generation of the tributylstannyl radical by mild UV irradiation in the presence of a triplet sensitizer (e.g. 4'-Methoxyacetophenone, A11162 ), allowing selective C-C bond forming reactions to be performed under free-radical conditions: Synlett, 287 (1993).
  2. Brings about the deoxygenation of amine oxides: Synthesis, 55 (1987), and the photo-desulfurization of 1,3-dithiole-2-thiones to tetrathiafulvalenes: J. Am. Chem. Soc., 98, 7440 (1976).
  3. Corcoran, E. B.; Williams, A. B.; Hanson, R. N. A Synthetic Method for Palladium-Catalyzed Stannylation at the 5-and 6-Benzo Positions of Indoles. Org. Lett. 2012, 14 (17), 4630-4633.
  4. Chun, J. H.; Pike, V. W. Regiospecific syntheses of functionalized diaryliodonium tosylates via [hydroxy (tosyloxy) iodo] arenes generated in situ from (diacetoxyiodo) arenes. J. Org. Chem. 2012, 77 (4), 1931-1938.