trans-Cinnamoyl chloride, 97%
trans-Cinnamoyl chloride, 97%
trans-Cinnamoyl chloride, 97%
trans-Cinnamoyl chloride, 97%
Thermo Scientific Chemicals

trans-Cinnamoyl chloride, 97%

CAS: 17082-09-6 | C9H7ClO | 166.604 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A12016.22
also known as A12016-22
Price (EUR)
68,09
Online Exclusive
80,10
Save 12,01 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
68,09
Online Exclusive
80,10
Save 12,01 (15%)
Each
Add to cart
Chemical Identifiers
CAS17082-09-6
IUPAC Name(2E)-3-phenylprop-2-enoyl chloride
Molecular FormulaC9H7ClO
InChI KeyWOGITNXCNOTRLK-VOTSOKGWSA-N
SMILESClC(=O)\C=C\C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormChunks or fused solid
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Melting Point (clear melt)31.0-40.0?C
Appearance (Color)White to pale cream
Assay (GC)≥96.0%
The derivative of this compound, trans-Cinnamoylimidazole is involved in the synthesis of trans-Cinnamoyl-Papain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The derivative of this compound, trans-Cinnamoylimidazole is involved in the synthesis of trans-Cinnamoyl-Papain.

Solubility
Reacts with water.

Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases. Moisture sensitive.
RUO – Research Use Only

General References:

  1. Lewis J. Brubacher.; Myron L. Bender. The Preparation and Properties of trans-Cinnamoyl-Papain. J. Am. Chem. Soc. 1966, 88 (24),5871-5880.
  2. Stefan Latza.; Dietmar Gansser.; Ralf G. Berger. Identification and Accumulation of 1-O-trans-Cinnamoyl-β-d-glucopyranose in Developing Strawberry Fruit (Fragaria ananassa Duch. Cv. Kent). J. Agric. Food Chem. 1996, 44 (6),1367-1370.