3-Chloroaniline, 99%
3-Chloroaniline, 99%
3-Chloroaniline, 99%
Thermo Scientific Chemicals

3-Chloroaniline, 99%

CAS: 108-42-9 | C6H6ClN | 127.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A12019.22
also known as A12019-22
Price (EUR)
27,90
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
27,90
Each
Chemical Identifiers
CAS108-42-9
IUPAC Name3-chloroaniline
Molecular FormulaC6H6ClN
InChI KeyPNPCRKVUWYDDST-UHFFFAOYSA-N
SMILESNC1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.5925-1.5955 @ 20?C
3-Chloroaniline was the only aniline compound to increase plasmaALT/GPT activity at 48 h. It is employed in the bioaugmentation of activated sludge.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloroaniline was the only aniline compound to increase plasmaALT/GPT activity at 48 h. It is employed in the bioaugmentation of activated sludge.

Solubility
Soluble in water. [6.8 g/L (20°C)].

Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases.
RUO – Research Use Only

General References:

  1. Nico Boon.; Johan Goris.; Paul De Vos.; Willy Verstraete.; Eva M. Top. Bioaugmentation of Activated Sludge by an Indigenous 3-Chloroaniline-Degrading Comamonas testosteroni Strain, I2gfp. Appl. Environ. Microbiol. 2000, 66 (7),2906-2913.
  2. Nico Boon.; Johan Goris.; Paul De Vos.; Willy Verstraete.; Eva M. Top. Genetic Diversity among 3-Chloroaniline- and Aniline-Degrading Strains of theComamonadaceae. Appl. Environ. Microbiol. 2001, 67 (3),1107-1115.