2-Isopropylphenol, 98+%
2-Isopropylphenol, 98+%
2-Isopropylphenol, 98+%
Thermo Scientific Chemicals

2-Isopropylphenol, 98+%

CAS: 88-69-7 | C9H12O | 136.194 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A12027.0B
also known as A12027-0B
Price (EUR)
149,00
Each
Quantity:
1000 g
Request bulk or custom format
Price (EUR)
149,00
Each
Specifications
Assay Percent Range98%
Beilstein1363322
CAS88-69-7
ChEBICHEBI:38506
Chemical Name or Material2-Isopropylphenol
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only