3-Fluoro-4-hydroxyphenylacetic acid, 98%
3-Fluoro-4-hydroxyphenylacetic acid, 98%
3-Fluoro-4-hydroxyphenylacetic acid, 98%
Thermo Scientific Chemicals

3-Fluoro-4-hydroxyphenylacetic acid, 98%

CAS: 458-09-3 | C8H7FO3 | 170.139 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A12114.03
also known as A12114-03
Price (EUR)
27,90
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
27,90
Each
Chemical Identifiers
CAS458-09-3
IUPAC Name2-(3-fluoro-4-hydroxyphenyl)acetate
Molecular FormulaC8H6FO3
InChI KeyYRFBZAHYMOSSGX-UHFFFAOYSA-M
SMILESOC1=CC=C(CC([O-])=O)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5% (Dry wt. basis)
Assay (HPLC)≥97.5%
Water Content (Karl Fischer Titration)<5.0%
Appearance (Color)White to cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only