Potassium permanganate, 98%
Potassium permanganate, 98%
Potassium permanganate, 98%
Thermo Scientific Chemicals

Potassium permanganate, 98%

CAS: 7722-64-7 | KMnO4 | 158.032 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 g
500 g
2500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A12170.0C
also known as A12170-0C
Price (EUR)
-
Quantity:
10,000 g
Request bulk or custom format
Chemical Identifiers
CAS7722-64-7
IUPAC Namepotassium manganesoylolate
Molecular FormulaKMnO4
InChI KeyVZJVWSHVAAUDKD-UHFFFAOYSA-N
SMILES[K+].[O-][Mn](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark purple to black
Assay (Iodometric Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
Potassium permanganate is an oxidizing agent, is used in the synthesis of organic compounds like ascorbic acid, chloramphenicol, saccharin, isonicotinic acid and pyrazinoic acid and in pharmaceutical intermediate. It is used to remove ferrous impurities from inorganic chemicals. It is also used in analytical chemistry as a reagent for redox titrations and to determine the kappa number of wood pulp. It is mostly used in water treatment and as a disinfectant. It also shows useful application as a chemiluminescence reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium permanganate is an oxidizing agent, is used in the synthesis of organic compounds like ascorbic acid, chloramphenicol, saccharin, isonicotinic acid and pyrazinoic acid and in pharmaceutical intermediate. It is used to remove ferrous impurities from inorganic chemicals. It is also used in analytical chemistry as a reagent for redox titrations and to determine the kappa number of wood pulp. It is mostly used in water treatment and as a disinfectant. It also shows useful application as a chemiluminescence reagent.

Solubility
Soluble in water.

Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong reducing agents, powdered metals, peroxides, zinc, copper, alcohols, hydrogen fluoride and acids.
RUO – Research Use Only

General References:

  1. The oxidizing power of permanganate can be varied according to the pH of the reaction medium, the use of co-reagents, phase-transfer catalysis, etc. For a comprehensive review of permanganate oxidations in organic chemistry, with particular emphasis on novel applications, see: Synthesis, 85, (1987).
  2. For phase-transfer oxidation, catalyzed by Tetra-n-butyl ammonium hydrogen sulfate, A14047, of unfunctionalized benzylic methylenes, or benzylic alcohols to carbonyls, see: Synthesis, 915 (1987). Similar transformations have been carried out in the presence of triethylamine: Synthesis, 293 (1989).
  3. For the use of (1-Hexadecyl) trimethyl ammonium bromide, A15235, as a surfactant in permanganate oxidations in micelles, see: Can. J. Chem., 67, 1381 (1989).
  4. Classical permanganate cleavage of olefins is difficult to control, and normally proceeds to the highest possible oxidation level. Adsorption of the reagent on moist alumina has been found to attenuate its activity and allow good yields of aldehydes to be obtained. This would appear to provide a possible alternative to ozonolysis for this conversion: J. Org. Chem., 58, 2918 (1993). For use in the cis-hydroxylation of olefins, see Dicyclohexano-18-crown-6, A15344.
  5. For phase-transfer assisted oxidation of primary amines to azo compounds, see: Bull. Soc. Chim. Belg., 102, 59 (1993).
  6. In combination with CuSO 4, has been used as a reagent for the heterogeneous oxidative cyclization of the primary position of a primary-secondary diol to the lactone: J. Chem. Soc., Chem. Commun., 634 (1988); Org, Synth. Coll., 9, 462 (1998):
  7. In combination with TMS chloride and benzyl triethylammonium chloride, generates a reagent for mild anti dichlorination of alkenes, conversion of epoxides to chlorohydrins, and selective oxidation of sulfides to sulfoxides: Tetrahedron Lett., 38, 2339 (1997).